Assay Details
Target Gene Details
Entrez Gene ID: | 29803 |
Gene Name: | replication initiator 1 |
Gene Aliases: |
AP4, RIP60, ZNF464, Zfp464 |
Location: |
Chr.7:150368228-150374044 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| REPIN1 | NM_001099695.1 | 3 | 2555 | NP_001093165.1 |
| NM_001099696.2 | 2 | 2357 | NP_001093166.1 | |
| NM_013400.3 | 4 | 2680 | NP_037532.2 | |
| NM_014374.3 | 1 | 2264 | NP_055189.2 | |
| XM_005249985.1 | 4 | 2575 | XP_005250042.1 | |
| XM_006715947.3 | 3 | 2589 | XP_006716010.1 | |
| XM_006715948.3 | 3 | 2609 | XP_006716011.1 | |
| XM_006715949.3 | 3 | 2494 | XP_006716012.1 | |
| XM_006715950.3 | 3 | 2586 | XP_006716013.1 | |
| XM_006715951.2 | 3 | 2515 | XP_006716014.1 | |
| XM_006715952.2 | 4 | 2683 | XP_006716015.1 | |
| XM_006715953.2 | 2 | 2347 | XP_006716016.1 | |
| XM_011516112.1 | 2 | 3337 | XP_011514414.1 | |
| XM_017012081.1 | 3 | 3137 | XP_016867570.1 | |
| XM_017012082.1 | 4 | 2743 | XP_016867571.1 | |
| AF201303.1 | 2 | 2320 | AAF26712.1 | |
| AJ245553.1 | 1 | 2264 | CAB53100.1 | |
| AK025356.1 | 1 | 2218 | ||
| AK226081.1 | 1 | 2269 | ||
| AL046625.1 | ||||
| BC000363.2 | 1 | 2246 | ||
| BC001760.1 | 1 | 2205 | AAH01760.1 | |
| CA449524.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034409 | Chr.7:150147550 - 150749983 on Build GRCh38 | Gain |
|
| nsv1015582 | Chr.7:150270897 - 150588016 on Build GRCh38 | Gain |
|
| nsv608990 | Chr.7:150264185 - 151010400 on Build GRCh38 | Gain |
|
| nsv521407 | Chr.7:150323206 - 150421502 on Build GRCh38 | Loss |
|
| dgv11718n54 | Chr.7:150338437 - 150418095 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map