Assay Details
Target Gene Details
Entrez Gene ID: | 146691 |
Gene Name: | target of myb1 like 2 membrane trafficking protein |
Gene Aliases: |
- |
Location: |
Chr.17:17843508-17972470 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOM1L2 | NM_001033551.2 | 14 | 1851 | NP_001028723.1 |
| NM_001082968.1 | 15 | 2001 | NP_001076437.1 | |
| NM_001288786.1 | 15 | 1929 | NP_001275715.1 | |
| NM_001288787.1 | 14 | 1866 | NP_001275716.1 | |
| NM_001288788.1 | 12 | 1647 | NP_001275717.1 | |
| NM_001288789.1 | 13 | 1644 | NP_001275718.1 | |
| XM_005256461.2 | 16 | 2022 | XP_005256518.1 | |
| XM_005256462.1 | 15 | 1889 | XP_005256519.1 | |
| XM_005256463.2 | 14 | 1875 | XP_005256520.1 | |
| XM_005256466.1 | 14 | 1730 | XP_005256523.1 | |
| XM_011523662.1 | 15 | 1874 | XP_011521964.1 | |
| XM_017024201.1 | 13 | 1742 | XP_016879690.1 | |
| AF467441.1 | AAL78338.1 | |||
| AK057308.1 | 9 | 1607 | BAB71421.1 | |
| AK124331.1 | 15 | 1929 | BAC85834.1 | |
| AK301944.1 | 14 | 1793 | ||
| BC051650.1 | 14 | 1791 | AAH51650.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833385 | Chr.17:17690723 - 17862264 on Build GRCh38 | Loss |
|
| nsv522668 | Chr.17:17716545 - 17849495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map