Assay Details
Target Gene Details
Entrez Gene ID: | 56948 |
Gene Name: | short chain dehydrogenase/reductase family 39U member 1 |
Gene Aliases: |
C14orf124, HCDI |
Location: |
Chr.14:24439766-24442905 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDR39U1 | NM_001290292.1 | NP_001277221.1 | ||
| NM_001290293.2 | 3 | 232 | NP_001277222.1 | |
| NM_001308075.1 | NP_001295004.1 | |||
| NM_020195.2 | NP_064580.2 | |||
| XM_011536975.1 | 3 | 205 | XP_011535277.1 | |
| XM_011536976.1 | XP_011535278.1 | |||
| AF226050.1 | AAF86950.1 | |||
| AK123513.1 | 2 | 290 | ||
| BC000989.2 | AAH00989.1 | |||
| BI256676.1 | ||||
| BX161384.1 | 2 | 183 | CAD61875.1 | |
| CN406892.1 | ||||
| CR995681.1 | ||||
| DB443637.1 | 3 | 199 | ||
| DT220226.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564061 | Chr.14:24441932 - 24476125 on Build GRCh38 | Loss |
|
| dgv1854n100 | Chr.14:24438841 - 24524460 on Build GRCh38 | Gain |
|
| nsv482171 | Chr.14:24439768 - 24442801 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map