Assay Details
Target Gene Details
Entrez Gene ID: | 157570 |
Gene Name: | establishment of sister chromatid cohesion N-acetyltransferase 2 |
Gene Aliases: |
2410004I17Rik, EFO2, RBS |
Location: |
Chr.8:27771949-27812404 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ESCO2 | NM_001017420.2 | 3 | 813 | NP_001017420.1 |
| XM_011544421.2 | 3 | 866 | XP_011542723.1 | |
| XM_011544422.2 | 3 | 814 | XP_011542724.1 | |
| AF306676.1 | 2 | 754 | ||
| AF306679.1 | 3 | 810 | ||
| AK124215.1 | 3 | 815 | ||
| AL832666.1 | 3 | 800 | ||
| AY882862.1 | 3 | 813 | AAX68677.1 | |
| BC034641.1 | 3 | 800 | AAH34641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524133 | Chr.8:27776547 - 27858791 on Build GRCh38 | Loss |
|
| nsv1017032 | Chr.8:27767011 - 27821659 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map