Assay Details
Target Gene Details
Entrez Gene ID: | 51507 |
Gene Name: | replication termination factor 2 domain containing 1 |
Gene Aliases: |
C20orf43, CDAO5, HSPC164, SHUJUN-3 |
Location: |
Chr.20:56468585-56518886 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTFDC1 | NM_001283035.1 | 5 | 537 | NP_001269964.1 |
| NM_001283036.1 | 4 | 447 | NP_001269965.1 | |
| NM_001283037.1 | 4 | 447 | NP_001269966.1 | |
| NM_016407.4 | 4 | 447 | NP_057491.2 | |
| XM_017027872.1 | 7 | 640 | XP_016883361.1 | |
| AF117231.1 | 4 | 395 | AAF17212.1 | |
| AF161513.1 | 4 | 404 | AAF29128.1 | |
| AF161518.1 | 4 | 373 | AAF29133.1 | |
| AF212244.1 | 4 | 384 | AAK14929.1 | |
| AK000481.1 | 4 | 377 | BAA91193.1 | |
| AK000586.1 | 3 | 267 | ||
| AK026118.1 | 4 | 375 | ||
| AK225135.1 | 4 | 365 | ||
| AK293662.1 | ||||
| AK302029.1 | 5 | 480 | ||
| AK314563.1 | 4 | 402 | ||
| BC003359.1 | 4 | 374 | AAH03359.1 | |
| BG468509.1 | 4 | 408 | ||
| BP307634.1 | 4 | 441 | ||
| BU537939.1 | 4 | 357 | ||
| CR983692.1 | 4 | 358 | ||
| DC397043.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv787e214 | Chr.20:56429038 - 56481532 on Build GRCh38 | Gain |
|
| esv3646189 | Chr.20:56430399 - 56481532 on Build GRCh38 | Loss |
|
| nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map