Assay Details
Target Gene Details
Entrez Gene ID: | 7068 |
Gene Name: | thyroid hormone receptor beta |
Gene Aliases: |
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2 |
Location: |
Chr.3:24117153-24495281 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THRB | NM_000461.4 | NP_000452.2 | ||
| NM_001128176.2 | NP_001121648.1 | |||
| NM_001128177.1 | NP_001121649.1 | |||
| NM_001252634.1 | NP_001239563.1 | |||
| XM_005265421.4 | XP_005265478.1 | |||
| XM_005265423.4 | XP_005265480.1 | |||
| XM_005265424.3 | XP_005265481.1 | |||
| XM_006713317.3 | 1 | 238 | XP_006713380.1 | |
| XM_006713318.3 | 1 | 238 | XP_006713381.1 | |
| XM_011534047.2 | 1 | 238 | XP_011532349.1 | |
| XM_011534048.2 | XP_011532350.1 | |||
| XM_011534049.2 | XP_011532351.1 | |||
| XM_011534050.2 | 1 | 238 | XP_011532352.1 | |
| XM_011534053.2 | XP_011532355.1 | |||
| XM_017007108.1 | 1 | 238 | XP_016862597.1 | |
| XM_017007109.1 | 1 | 238 | XP_016862598.1 | |
| XM_017007110.1 | 1 | 238 | XP_016862599.1 | |
| XM_017007113.1 | 1 | 238 | XP_016862602.1 | |
| XM_017007114.1 | 1 | 238 | XP_016862603.1 | |
| XM_017007115.1 | XP_016862604.1 | |||
| AK096628.1 | ||||
| AY286466.1 | AAQ23705.1 | |||
| AY286467.1 | AAQ23706.1 | |||
| AY286470.1 | AAQ23709.1 | |||
| BP313943.1 | ||||
| BQ879477.1 | ||||
| DA218319.1 | ||||
| M26747.1 | AAA35677.1 | |||
| X04707.1 | CAA28412.1 |
Target Gene Details
Entrez Gene ID: | 644990 |
Gene Name: | THRB antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:24494087-24500011 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THRB-AS1 | NR_046244.1 | |||
| AK130231.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012975 | Chr.3:24459215 - 24574153 on Build GRCh38 | Gain |
|
| nsv999384 | Chr.3:24483854 - 24615382 on Build GRCh38 | Gain |
|
| dgv74n68 | Chr.3:24424371 - 24599102 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map