Assay Details
Target Gene Details
Entrez Gene ID: | 91181 |
Gene Name: | nucleoporin 210 like |
Gene Aliases: |
- |
Location: |
Chr.1:153992582-154155116 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUP210L | NM_001159484.1 | 35 | 4905 | NP_001152956.1 |
| NM_207308.2 | 35 | 4905 | NP_997191.2 | |
| XM_011510122.1 | 34 | 4736 | XP_011508424.1 | |
| XM_011510123.1 | 34 | 4729 | XP_011508425.1 | |
| XM_011510124.1 | 35 | 4888 | XP_011508426.1 | |
| XM_017002788.1 | 35 | 4888 | XP_016858277.1 | |
| XM_017002789.1 | 35 | 4888 | XP_016858278.1 | |
| AK125924.1 | 31 | 4167 | BAC86345.1 | |
| AK128108.1 | 25 | 3484 | BAC87279.1 | |
| AL133633.1 | 13 | 1679 | CAB63757.1 | |
| AL833964.1 | 19 | 2412 | CAD38812.1 | |
| BX648132.1 | 23 | 3093 |
More Information
Set Membership: |
|

Genomic Map