Assay Details
Target Gene Details
Entrez Gene ID: | 6891 |
Gene Name: | transporter 2, ATP binding cassette subfamily B member |
Gene Aliases: |
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11 |
Location: |
Chr.6:32821833-32838823 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAP2 | NM_000544.3 | 12 | 2295 | NP_000535.3 |
| NM_001290043.1 | 12 | 2348 | NP_001276972.1 | |
| NM_018833.2 | NP_061313.2 | |||
| AB208953.1 | 12 | 2242 | BAD92190.1 | |
| AF078671.1 | AAD31384.1 | |||
| AF105151.1 | AAD12059.1 | |||
| AK222823.1 | 11 | 2204 | BAD96543.1 | |
| AK223300.1 | 12 | 2255 | BAD97020.1 | |
| AK225657.1 | ||||
| AK225693.1 | 12 | 2255 | ||
| AK292963.1 | 12 | 2278 | ||
| M74447.1 | 12 | 2269 | AAA59841.1 | |
| M84748.1 | 12 | 2201 | ||
| U07844.1 | 11 | 1983 | AAA79901.1 | |
| Z22936.1 | 12 | 2249 | CAA80523.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1073969 | Chr.6:31456422 - 33480523 on Build GRCh38 | Deletion |
|
| nsv1112900 | Chr.6:31455323 - 33480423 on Build GRCh38 | Deletion |
|
| nsv1119076 | Chr.6:32595823 - 33480623 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map