Assay Details
Target Gene Details
Entrez Gene ID: | 388531 |
Gene Name: | regulator of G-protein signaling 9 binding protein |
Gene Aliases: |
PERRS, R9AP, RGS9 |
Location: |
Chr.19:32675407-32678300 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGS9BP | NM_207391.2 | 1 | 1831 | NP_997274.2 |
| AK124499.1 | 1 | 1831 | BAC85865.1 | |
| AK127646.1 | 1 | 1388 | BAC87072.1 |
More Information
Set Membership: |
|

Genomic Map