Assay Details
Target Gene Details
Entrez Gene ID: | 93517 |
Gene Name: | short chain dehydrogenase/reductase family 42E, member 1 |
Gene Aliases: |
HSPC105 |
Location: |
Chr.16:81988855-82011488 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDR42E1 | NM_145168.2 | NP_660151.2 | ||
| XM_005256257.3 | XP_005256314.1 | |||
| AK026740.1 | ||||
| AK315652.1 | ||||
| BC019670.1 | AAH19670.1 | |||
| BX482154.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660861 | Chr.16:82010102 - 82015982 on Build GRCh38 | Deletion |
|
| esv3639348 | Chr.16:82010110 - 82016025 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs112716404] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map