Assay Details
Target Gene Details
Entrez Gene ID: | 266743 |
Gene Name: | neuronal PAS domain protein 4 |
Gene Aliases: |
Le-PAS, NXF, PASD10, bHLHe79 |
Location: |
Chr.11:66409158-66426707 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPAS4 | NM_001318804.1 | 6 | 2193 | NP_001305733.1 |
| NM_178864.3 | 7 | 2303 | NP_849195.2 | |
| XM_017017537.1 | 9 | 3742 | XP_016873026.1 | |
| XM_017017538.1 | 3 | 1455 | XP_016873027.1 | |
| AB049469.1 | 7 | 2228 | BAC19830.1 | |
| AK094025.1 | 4 | 1608 | BAC04271.1 | |
| AK096253.1 | 7 | 2303 | BAC04738.1 | |
| BC105001.1 | 7 | 2216 | AAI05002.1 | |
| BC105003.1 | 7 | 2216 | AAI05004.1 | |
| BC143630.1 | 6 | 2106 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832193 | Chr.11:66352868 - 66511334 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map