Assay Details
Target Gene Details
Entrez Gene ID: | 55226 |
Gene Name: | N-acetyltransferase 10 |
Gene Aliases: |
ALP, NET43 |
Location: |
Chr.11:34105564-34146911 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAT10 | NM_001144030.1 | NP_001137502.1 | ||
| NM_024662.2 | 2 | 303 | NP_078938.2 | |
| AB051496.1 | 2 | 224 | ||
| AK001636.1 | 2 | 158 | BAA91800.1 | |
| AK022241.1 | 2 | 265 | BAB13995.1 | |
| AK225341.1 | 2 | 265 | ||
| AK294044.1 | ||||
| AL136882.1 | 2 | 160 | CAB66816.1 | |
| BC020205.1 | 2 | 125 | ||
| BC035558.1 | 2 | 237 | AAH35558.1 | |
| CN336530.1 | 2 | 305 |
More Information
Set Membership: |
|

Genomic Map