Assay Details
Target Gene Details
Entrez Gene ID: | 137362 |
Gene Name: | glutamic-oxaloacetic transaminase 1-like 1 |
Gene Aliases: |
- |
Location: |
Chr.8:37933548-37940502 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOT1L1 | NM_152413.2 | NP_689626.2 | ||
| XM_005273399.3 | XP_005273456.1 | |||
| XM_006716285.3 | XP_006716348.1 | |||
| XM_011544391.2 | XP_011542693.1 | |||
| BC029504.1 | AAH29504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610947 | Chr.8:37935047 - 38054626 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map