Assay Details
Target Gene Details
Entrez Gene ID: | 25891 |
Gene Name: | peptidase domain containing associated with muscle regeneration 1 |
Gene Aliases: |
DKFZP586H2123, FP938, RAMP |
Location: |
Chr.11:35431827-35530300 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAMR1 | NM_001001991.2 | 11 | 2365 | NP_001001991.1 |
| NM_001282675.1 | 13 | 2355 | NP_001269604.1 | |
| NM_001282676.1 | 9 | 2032 | NP_001269605.1 | |
| NM_015430.3 | 12 | 2416 | NP_056245.2 | |
| AF370388.1 | 10 | 1854 | AAQ15224.1 | |
| AK027841.1 | 12 | 2016 | BAB55404.1 | |
| AK297092.1 | 13 | 2356 | ||
| AK300185.1 | 9 | 1617 | ||
| AK316188.1 | 9 | 1604 | ||
| AL050214.1 | 1 | 303 | CAB43317.1 | |
| AL832391.1 | 11 | 1931 | CAI46203.1 | |
| AY358346.1 | 11 | 1944 | AAQ88712.1 | |
| BC089434.1 | 12 | 2008 | AAH89434.1 | |
| BX640676.1 | 11 | 2365 | CAE45808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759819 | Chr.11:35297773 - 35458974 on Build GRCh38 | Gain |
|
| nsv1042014 | Chr.11:34919190 - 37195907 on Build GRCh38 | Loss |
|
| nsv832120 | Chr.11:35255863 - 35468756 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map