Assay Details
Target Gene Details
Entrez Gene ID: | 51290 |
Gene Name: | ERGIC and golgi 2 |
Gene Aliases: |
CDA14, Erv41, PTX1, cd002 |
Location: |
Chr.12:29336544-29381234 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERGIC2 | NM_016570.2 | 12 | 1058 | NP_057654.2 |
| AF183410.1 | 12 | 1012 | AAG09679.1 | |
| AF216751.1 | 12 | 1016 | AAF61208.1 | |
| AF302767.1 | 12 | 1011 | AAK77355.1 | |
| AK074520.1 | 12 | 1033 | BAC11037.1 | |
| AK222799.1 | 12 | 998 | BAD96519.1 | |
| AL834128.2 | 12 | 999 | CAD89902.1 | |
| BC000887.1 | 12 | 1011 | AAH00887.2 | |
| BC107794.1 | 12 | 1015 | AAI07795.1 |
More Information
Set Membership: |
|

Genomic Map