Assay Details
Target Gene Details
Entrez Gene ID: | 80709 |
Gene Name: | AT-hook transcription factor |
Gene Aliases: |
- |
Location: |
Chr.9:114334149-114398498 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12859n54 | Chr.9:114193988 - 114696195 on Build GRCh38 | Gain |
|
| nsv1161903 | Chr.9:114340988 - 114388671 on Build GRCh38 | Deletion |
|
| dgv7706n100 | Chr.9:114182745 - 114697870 on Build GRCh38 | Gain |
|
| nsv509319 | Chr.9:114297186 - 114380605 on Build GRCh38 | Insertion |
|
| dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication |
|
| nsv615233 | Chr.9:114167820 - 114486199 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map