Assay Details
Target Gene Details
Entrez Gene ID: | 27 |
Gene Name: | ABL proto-oncogene 2, non-receptor tyrosine kinase |
Gene Aliases: |
ABLL, ARG |
Location: |
Chr.1:179099327-179229693 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABL2 | NM_001136000.2 | NP_001129472.1 | ||
| NM_001168236.1 | NP_001161708.1 | |||
| NM_001168237.1 | NP_001161709.1 | |||
| NM_001168238.1 | NP_001161710.1 | |||
| NM_001168239.1 | NP_001161711.1 | |||
| NM_005158.4 | NP_005149.4 | |||
| NM_007314.3 | NP_009298.1 | |||
| XM_005245088.2 | XP_005245145.1 | |||
| XM_017001035.1 | XP_016856524.1 | |||
| BC065912.1 | AAH65912.1 | |||
| BX538317.1 | CAD98092.1 | |||
| FJ542283.1 | ||||
| FJ542284.1 | ||||
| FJ542285.1 | ||||
| FJ542286.1 | ||||
| M35296.1 | AAA35553.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002511 | Chr.1:179108779 - 179120707 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map