Assay Details
Target Gene Details
Entrez Gene ID: | 6793 |
Gene Name: | serine/threonine kinase 10 |
Gene Aliases: |
LOK, PRO2729 |
Location: |
Chr.5:172042070-172188342 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK10 | NM_005990.3 | 19 | 4242 | NP_005981.3 |
| XM_017009788.1 | 18 | 3796 | XP_016865277.1 | |
| AB015718.1 | 19 | 3992 | BAA35073.1 | |
| AF119894.1 | 1 | 939 | AAF69648.1 | |
| AK022960.1 | 11 | 2815 | ||
| AK092814.1 | 2 | 1448 | ||
| AL133081.1 | 4 | 1503 | CAB61400.1 | |
| BC070077.1 | 19 | 4156 | AAH70077.1 |
More Information
Set Membership: |
|

Genomic Map