Assay Details
Target Gene Details
Entrez Gene ID: | 25896 |
Gene Name: | integrator complex subunit 7 |
Gene Aliases: |
C1orf73, INT7 |
Location: |
Chr.1:211940328-212035660 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INTS7 | NM_001199809.1 | 8 | 1120 | NP_001186738.1 |
| NM_001199811.1 | 9 | 1267 | NP_001186740.1 | |
| NM_001199812.1 | 9 | 1267 | NP_001186741.1 | |
| NM_015434.3 | 9 | 1267 | NP_056249.1 | |
| NR_037667.1 | 10 | 1301 | ||
| XM_011509396.2 | 9 | 1152 | XP_011507698.1 | |
| XM_017000962.1 | 9 | 1152 | XP_016856451.1 | |
| AK001363.1 | 9 | 1127 | BAA91650.1 | |
| AK001598.1 | 9 | 1149 | BAA91779.1 | |
| AK022509.1 | 9 | 1127 | BAB14067.1 | |
| AK297225.1 | 8 | 999 | ||
| AL133049.2 | 4 | 436 | CAB61376.1 | |
| BC020523.2 | 9 | 1143 | AAH20523.1 | |
| BC030716.1 | 9 | 1122 | AAH30716.1 | |
| BC033918.1 | 9 | 1126 | AAH33918.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3588741 | Chr.1:211945914 - 212035544 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map