Assay Details
Target Gene Details
Entrez Gene ID: | 10588 |
Gene Name: | 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase) |
Gene Aliases: |
HsT19268 |
Location: |
Chr.15:79843547-79897285 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTHFS | NM_001199758.1 | 3 | 513 | NP_001186687.1 |
| NM_006441.3 | 3 | 566 | NP_006432.1 | |
| NR_037654.1 | 3 | 595 | ||
| AK054972.1 | 1 | 1871 | ||
| AL110101.1 | 3 | 512 | ||
| BC012417.1 | 3 | 476 | AAH12417.1 | |
| BC019921.1 | 3 | 566 | AAH19921.1 | |
| BM450865.1 | 3 | 583 | ||
| CB995322.1 | 3 | 539 | ||
| CK905549.1 | 1 | 357 | ||
| L38928.1 | 3 | 518 | AAC41945.1 |
Target Gene Details
Entrez Gene ID: | 100528021 |
Gene Name: | ST20-MTHFS readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:79843547-79923106 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST20-MTHFS | NM_001199760.1 | 4 | 512 | NP_001186689.1 |
| CK002935.1 | 4 | 516 | ||
| DB495490.1 | 2 | 427 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833065 | Chr.15:79760661 - 79982480 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map