Assay Details
Target Gene Details
Entrez Gene ID: | 5139 |
Gene Name: | phosphodiesterase 3A |
Gene Aliases: |
CGI-PDE, CGI-PDE A, CGI-PDE-A, HTNB |
Location: |
Chr.12:20369245-20684107 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE3A | NM_000921.4 | 1 | 460 | NP_000912.3 |
| XM_017019420.1 | 1 | 441 | XP_016874909.1 | |
| AJ005036.1 | 1 | 442 | CAA06304.1 | |
| BC117369.1 | 1 | 460 | ||
| BC117371.1 | 1 | 460 | ||
| M91667.2 | 1 | 449 | AAA35912.2 | |
| U36798.1 | 1 | 420 | AAB18673.1 |
More Information
Set Membership: |
|

Genomic Map