Assay Details
Target Gene Details
Entrez Gene ID: | 6427 |
Gene Name: | serine and arginine rich splicing factor 2 |
Gene Aliases: |
PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b |
Location: |
Chr.17:76734115-76737431 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF2 | NM_001195427.1 | 3 | 1064 | NP_001182356.1 |
| NM_003016.4 | 2 | 2059 | NP_003007.2 | |
| NR_036608.1 | 4 | 1168 | ||
| XM_017024942.1 | 3 | 1801 | XP_016880431.1 | |
| AK092489.1 | 4 | 924 | BAC03903.1 | |
| AK093626.1 | 3 | 1630 | BAC04206.1 | |
| AK097069.1 | 3 | 1849 | ||
| AK098209.1 | 3 | 1950 | ||
| AK123712.1 | 2 | 1881 | ||
| AK124792.1 | 3 | 1027 | ||
| AK126432.1 | 3 | 1438 | ||
| AK223252.1 | 3 | 983 | BAD96972.1 | |
| AK297813.1 | 3 | 948 | ||
| BC000339.2 | 3 | 964 | AAH00339.1 | |
| BC001303.2 | 3 | 961 | AAH01303.1 | |
| BC066958.1 | AAH66958.1 | |||
| BC070086.1 | 2 | 1979 | AAH70086.1 | |
| EB388696.1 | 4 | 581 | ||
| M90104.1 | 3 | 968 | AAA60306.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833550 | Chr.17:76712038 - 76886290 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map