Assay Details
Target Gene Details
Entrez Gene ID: | 4715 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit B9 |
Gene Aliases: |
B22, CI-B22, LYRM3, UQOR22 |
Location: |
Chr.8:124539102-124549986 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFB9 | NM_001278645.1 | NP_001265574.1 | ||
| NM_001278646.1 | NP_001265575.1 | |||
| NM_001311168.1 | NP_001298097.1 | |||
| NM_005005.2 | NP_004996.1 | |||
| AF044956.1 | AAD42057.1 | |||
| AF067168.1 | AAD32452.1 | |||
| AF261090.1 | AAF99683.1 | |||
| AI907018.1 | ||||
| AK026538.1 | ||||
| AK302286.1 | ||||
| AK313432.1 | ||||
| AL136714.1 | CAI46222.1 | |||
| BC007672.2 | AAH07672.1 | |||
| BG703246.1 | ||||
| BQ685604.1 | ||||
| CN410734.1 | ||||
| DB504123.1 | ||||
| S82655.1 | AAB46797.2 |
More Information
Set Membership: |
|

Genomic Map