Assay Details
Target Gene Details
Entrez Gene ID: | 1757 |
Gene Name: | sarcosine dehydrogenase |
Gene Aliases: |
BPR-2, DMGDHL1, SAR, SARD, SDH |
Location: |
Chr.9:133663074-133739958 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SARDH | NM_001134707.1 | 21 | 3130 | NP_001128179.1 |
| NM_007101.3 | 21 | 3289 | NP_009032.2 | |
| XM_017014367.1 | XP_016869856.1 | |||
| AF095735.2 | 21 | 3254 | AAD53398.2 | |
| AF129265.1 | 17 | 2281 | AAD33412.1 | |
| AK093794.1 | 9 | 2180 | ||
| AK223402.1 | 21 | 3176 | BAD97122.1 | |
| AK298348.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv998446 | Chr.9:133660557 - 133670710 on Build GRCh38 | Deletion |
|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| dgv12937n54 | Chr.9:133660184 - 133669269 on Build GRCh38 | Gain |
|
| nsv951800 | Chr.9:133627979 - 133696678 on Build GRCh38 | Deletion |
|
| esv3576711 | Chr.9:133582804 - 133778331 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map