Assay Details
Target Gene Details
Entrez Gene ID: | 337867 |
Gene Name: | UBA domain containing 2 |
Gene Aliases: |
PHGDHL1 |
Location: |
Chr.13:99200425-99386499 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBAC2 | NM_001144072.1 | NP_001137544.1 | ||
| NM_177967.3 | NP_808882.1 | |||
| NR_026644.1 | ||||
| XM_006719948.3 | XP_006720011.1 | |||
| XM_011521082.2 | XP_011519384.1 | |||
| XM_011521083.2 | XP_011519385.1 | |||
| XM_011521084.2 | XP_011519386.1 | |||
| AK054563.1 | BAB70757.1 | |||
| AK075419.1 | BAC11609.1 | |||
| AK075516.1 | BAC11665.1 | |||
| AK124404.1 | ||||
| AK129861.1 | ||||
| AK290342.1 | ||||
| AK300908.1 | ||||
| BC121138.1 | ||||
| BC121139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892433 | Chr.13:98843569 - 99243711 on Build GRCh38 | Gain |
|
| nsv1050094 | Chr.13:98974062 - 99323969 on Build GRCh38 | Gain |
|
| esv3582749 | Chr.13:99137911 - 99253291 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map