Assay Details
Target Gene Details
Entrez Gene ID: | 833 |
Gene Name: | cysteinyl-tRNA synthetase |
Gene Aliases: |
CARS1, CYSRS, MGC:11246 |
Location: |
Chr.11:3000922-3057451 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARS | NM_001014437.2 | 23 | 2682 | NP_001014437.1 |
| NM_001194997.1 | 23 | 2690 | NP_001181926.1 | |
| NM_001751.5 | 22 | 2433 | NP_001742.1 | |
| NM_139273.3 | 22 | 2441 | NP_644802.1 | |
| NR_036542.1 | 23 | 2729 | ||
| XM_006718341.3 | 22 | 2456 | XP_006718404.1 | |
| XM_017018389.1 | 22 | 2464 | XP_016873878.1 | |
| XM_017018390.1 | 23 | 2704 | XP_016873879.1 | |
| XM_017018391.1 | 20 | 2680 | XP_016873880.1 | |
| XM_017018392.1 | 22 | 2626 | XP_016873881.1 | |
| AB208814.1 | 19 | 5650 | BAD92051.1 | |
| AF288206.1 | AAG00578.1 | |||
| AF288207.1 | AAG00579.1 | |||
| AI432096.1 | 1 | 118 | ||
| AI608729.1 | 1 | 95 | ||
| AK096313.1 | 23 | 2696 | ||
| AK125503.1 | 5 | 2589 | ||
| AK126135.1 | 5 | 2372 | ||
| AK222614.1 | 22 | 2382 | BAD96334.1 | |
| AK295371.1 | 22 | 2332 | ||
| AK296763.1 | 23 | 2555 | ||
| AK298514.1 | 23 | 2657 | ||
| AK302644.1 | 23 | 2649 | ||
| BC002880.2 | 22 | 2419 | AAH02880.1 | |
| BX647906.1 | 23 | 2656 | CAI46108.1 | |
| L06845.1 | 20 | 2227 | AAA73901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv8766 | Chr.11:2971449 - 3205956 on Build GRCh38 | Loss |
|
| esv3625132 | Chr.11:2983355 - 3019578 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map