Assay Details
Target Gene Details
Entrez Gene ID: | 113802 |
Gene Name: | HEN1 methyltransferase homolog 1 |
Gene Aliases: |
C1orf59, HEN1 |
Location: |
Chr.1:108648288-108661526 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HENMT1 | NM_001102592.1 | NP_001096062.1 | ||
| NM_144584.2 | NP_653185.2 | |||
| XM_005270411.1 | XP_005270468.1 | |||
| XM_017000179.1 | XP_016855668.1 | |||
| AK055087.1 | BAB70852.1 | |||
| AK310152.1 | ||||
| BC012198.1 | AAH12198.1 | |||
| BC088366.1 | AAH88366.1 |
More Information
Set Membership: |
|

Genomic Map