Assay Details
Target Gene Details
Entrez Gene ID: | 9901 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 3 |
Gene Aliases: |
ARHGAP14, MEGAP, SRGAP2, WRP |
Location: |
Chr.3:8980591-9362979 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP3 | NM_001033117.2 | 22 | 5933 | NP_001028289.1 |
| NM_014850.3 | 22 | 6005 | NP_055665.1 | |
| XM_011534295.1 | 23 | 6020 | XP_011532597.1 | |
| XM_011534296.1 | 24 | 6137 | XP_011532598.1 | |
| XM_011534301.2 | 20 | 7849 | XP_011532603.1 | |
| XM_011534303.1 | 18 | 4745 | XP_011532605.1 | |
| XM_017007574.1 | 22 | 6343 | XP_016863063.1 | |
| XM_017007575.1 | 22 | 6271 | XP_016863064.1 | |
| XM_017007576.1 | 24 | 5512 | XP_016863065.1 | |
| XM_017007577.1 | 24 | 5451 | XP_016863066.1 | |
| XM_017007578.1 | 22 | 6371 | XP_016863067.1 | |
| XM_017007579.1 | 21 | 6011 | XP_016863068.1 | |
| AB007871.2 | 22 | 5157 | BAA24841.2 | |
| AF427144.1 | 22 | 5373 | AAN07095.1 | |
| AF464189.1 | 22 | 5461 | AAN57784.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3689 | Chr.3:8967425 - 8993205 on Build GRCh38 | Deletion |
|
| nsv518352 | Chr.3:8905994 - 9089915 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map