Assay Details
Target Gene Details
Entrez Gene ID: | 6777 |
Gene Name: | signal transducer and activator of transcription 5B |
Gene Aliases: |
STAT5 |
Location: |
Chr.17:42199177-42276460 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAT5B | NM_012448.3 | 19 | 4517 | NP_036580.2 |
| XM_017024977.1 | 18 | 4336 | XP_016880466.1 | |
| AB208920.1 | 19 | 4496 | BAD92157.1 | |
| AL080218.1 | 1 | 1023 | ||
| BC065227.1 | 19 | 4517 | AAH65227.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map