Assay Details
Target Gene Details
Entrez Gene ID: | 10307 |
Gene Name: | amyloid beta precursor protein binding family B member 3 |
Gene Aliases: |
FE65L2, SRA |
Location: |
Chr.5:140558268-140564604 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APBB3 | NM_006051.3 | NP_006042.3 | ||
| NM_133172.2 | NP_573418.2 | |||
| NM_133173.2 | NP_573419.2 | |||
| NM_133174.2 | NP_573420.2 | |||
| AB018247.1 | BAA35188.1 | |||
| AB049618.1 | BAB40806.1 | |||
| AB061224.1 | BAB71767.1 | |||
| AB079687.1 | BAC56948.1 | |||
| AB094990.1 | BAC22578.1 | |||
| AB209839.1 | BAD93076.1 | |||
| AF224708.1 | AAF65172.1 | |||
| AF224709.1 | AAF65173.1 | |||
| AF224710.1 | AAF65174.1 | |||
| AF224711.1 | AAF65175.1 | |||
| AK075280.1 | ||||
| AK125244.1 | ||||
| AK304824.1 | ||||
| AK307170.1 | 3 | 603 | ||
| BC013158.1 | AAH13158.1 | |||
| BC125187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462462 | Chr.5:140556654 - 140626201 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map