Assay Details
Target Gene Details
Entrez Gene ID: | 130367 |
Gene Name: | sphingosine-1-phosphate phosphatase 2 |
Gene Aliases: |
SPP2, SPPase2 |
Location: |
Chr.2:222424052-222562622 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGPP2 | NM_001320833.1 | 1 | 127 | NP_001307762.1 |
| NM_001320834.1 | NP_001307763.1 | |||
| NM_152386.3 | 1 | 127 | NP_689599.2 | |
| AF542512.1 | 1 | 31 | AAN28731.1 | |
| AK096323.1 | BAC04762.1 | |||
| AK314322.1 | 1 | 127 | ||
| BC106904.1 | AAI06905.1 | |||
| BC106905.1 | 1 | 24 | AAI06906.1 | |
| BC134342.1 | 1 | 31 |
More Information
Set Membership: |
|

Genomic Map