Assay Details
Target Gene Details
Entrez Gene ID: | 6599 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1 |
Gene Aliases: |
BAF155, CRACC1, Rsc8, SRG3, SWI3 |
Location: |
Chr.3:47585888-47781915 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMARCC1 | NM_003074.3 | 26 | 2933 | NP_003065.3 |
| XM_011534034.1 | 26 | 2664 | XP_011532336.1 | |
| AK302412.1 | 13 | 1377 | ||
| AK302541.1 | 27 | 2708 | ||
| BC050564.1 | 26 | 2862 | AAH50564.1 | |
| BC113465.1 | 26 | 2881 | ||
| BC117213.1 | 26 | 2881 | ||
| U66615.1 | 26 | 2866 | AAC50693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map