Assay Details
Target Gene Details
Entrez Gene ID: | 64768 |
Gene Name: | inositol-pentakisphosphate 2-kinase |
Gene Aliases: |
C9orf12, INSP5K2, IP5K, IPK1, bA476B13.1 |
Location: |
Chr.9:92613184-92670265 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPPK | NM_022755.5 | NP_073592.1 | ||
| XM_017015041.1 | XP_016870530.1 | |||
| AF351202.1 | AAK69692.1 | |||
| AF520811.1 | AAM75353.1 | |||
| AK023225.1 | BAB14476.1 | |||
| AK024267.1 | BAB14866.1 | |||
| AK096392.1 | ||||
| AK123694.1 | ||||
| BC026154.2 | AAH26154.1 |
Target Gene Details
Entrez Gene ID: | 100128361 |
Gene Name: | uncharacterized LOC100128361 |
Gene Aliases: |
- |
Location: |
Chr.9:92618050-92620533 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100128361 | NR_036505.1 | 1 | 1462 | |
| AK094300.1 | 1 | 1462 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950893 | Chr.9:92619219 - 92621818 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map