Assay Details
Target Gene Details
Entrez Gene ID: | 5289 |
Gene Name: | phosphatidylinositol 3-kinase catalytic subunit type 3 |
Gene Aliases: |
VPS34, Vps34, hVps34 |
Location: |
Chr.18:41955196-42084606 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3C3 | NM_001308020.1 | 1 | 98 | NP_001294949.1 |
| NM_002647.3 | 1 | 98 | NP_002638.2 | |
| XM_011526031.2 | 1 | 100 | XP_011524333.1 | |
| AK298517.1 | ||||
| AK308364.1 | ||||
| BC033004.2 | AAH33004.1 | |||
| BC053651.1 | AAH53651.1 | |||
| BI562123.1 | 1 | 92 | ||
| DB450556.1 | 1 | 99 | ||
| Z46973.1 | 1 | 51 | CAA87094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893097 | Chr.18:41680652 - 42201288 on Build GRCh38 | Gain |
|
| esv34153 | Chr.18:41818803 - 42292300 on Build GRCh38 | Loss |
|
| esv3642321 | Chr.18:41315081 - 42116859 on Build GRCh38 | Gain |
|
| esv3642342 | Chr.18:41818170 - 41981419 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map