Assay Details
Target Gene Details
Entrez Gene ID: | 56850 |
Gene Name: | GRIP1 associated protein 1 |
Gene Aliases: |
GRASP-1 |
Location: |
Chr.X:48973720-49002264 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIPAP1 | NM_020137.4 | NP_064522.3 | ||
| XM_011543935.1 | XP_011542237.1 | |||
| XM_011543936.1 | XP_011542238.1 | |||
| XM_011543937.1 | XP_011542239.1 | |||
| XM_017029656.1 | XP_016885145.1 | |||
| XM_017029657.1 | XP_016885146.1 | |||
| XM_017029658.1 | XP_016885147.1 | |||
| XM_017029659.1 | XP_016885148.1 | |||
| XM_017029660.1 | XP_016885149.1 | |||
| XM_017029661.1 | XP_016885150.1 | |||
| AB032993.1 | BAA86481.1 | |||
| AL136847.1 | ||||
| BC001522.2 | AAH01522.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528292 | Chr.X:48433286 - 49274368 on Build GRCh38 | Gain |
|
| esv32909 | Chr.X:48003374 - 49285646 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map