Assay Details
Target Gene Details
Entrez Gene ID: | 221395 |
Gene Name: | adhesion G protein-coupled receptor F5 |
Gene Aliases: |
GPR116, KPG_001 |
Location: |
Chr.6:46852505-46954954 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRF5 | NM_001098518.1 | 15 | 2403 | NP_001091988.1 |
| NM_015234.4 | 15 | 2343 | NP_056049.4 | |
| XM_005248892.2 | 16 | 2523 | XP_005248949.1 | |
| XM_005248893.3 | 16 | 2479 | XP_005248950.1 | |
| XM_005248894.3 | 16 | 2523 | XP_005248951.1 | |
| XM_005248895.3 | 16 | 2523 | XP_005248952.1 | |
| XM_011514353.2 | 15 | 2403 | XP_011512655.1 | |
| XM_011514354.2 | 17 | 2546 | XP_011512656.1 | |
| XM_017010418.1 | 16 | 2479 | XP_016865907.1 | |
| XM_017010419.1 | 15 | 2359 | XP_016865908.1 | |
| XM_017010420.1 | 15 | 2403 | XP_016865909.1 | |
| XM_017010421.1 | 15 | 2359 | XP_016865910.1 | |
| XM_017010422.1 | 15 | 2319 | XP_016865911.1 | |
| XM_017010423.1 | 16 | 2426 | XP_016865912.1 | |
| XM_017010424.1 | 16 | 2426 | XP_016865913.1 | |
| AB018301.2 | 15 | 2341 | ||
| AB031421.1 | 14 | 2158 | BAD83582.1 | |
| AK289503.1 | 15 | 2341 | ||
| AK296948.1 | 5 | 726 | ||
| AK300380.1 | 4 | 914 | ||
| AL832125.1 | 13 | 1884 | ||
| AY140958.1 | 14 | 2111 | AAN46672.1 | |
| AY498875.1 | 15 | 2202 | AAS21061.1 | |
| BC050087.1 | 2 | 227 | AAH50087.1 | |
| BC066121.1 | 15 | 2181 | AAH66121.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830654 | Chr.6:46755161 - 46938169 on Build GRCh38 | Gain |
|
| nsv428147 | Chr.6:46732445 - 46923866 on Build GRCh38 | Gain |
|
| esv24845 | Chr.6:46827810 - 46864239 on Build GRCh38 | Gain |
|
| nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map