Assay Details
Target Gene Details
Entrez Gene ID: | 79574 |
Gene Name: | EPS8 like 3 |
Gene Aliases: |
EPS8R3 |
Location: |
Chr.1:109750080-109764022 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPS8L3 | NM_001319952.1 | NP_001306881.1 | ||
| NM_024526.3 | NP_078802.2 | |||
| NM_133181.3 | NP_573444.2 | |||
| NM_139053.2 | NP_620641.1 | |||
| XM_011542132.1 | XP_011540434.1 | |||
| XM_011542133.1 | XP_011540435.1 | |||
| XM_011542134.2 | XP_011540436.1 | |||
| XM_011542135.2 | XP_011540437.1 | |||
| XM_017002327.1 | XP_016857816.1 | |||
| XM_017002328.1 | XP_016857817.1 | |||
| XM_017002329.1 | XP_016857818.1 | |||
| AK025175.1 | BAB15081.1 | |||
| AK225235.1 | ||||
| AK290261.1 | ||||
| AK292198.1 | ||||
| AK307932.1 | ||||
| AK308196.1 | ||||
| AK310979.1 | ||||
| AW937714.1 | ||||
| AY074930.1 | AAL76119.1 | |||
| BC012926.1 | AAH12926.1 | |||
| BP419077.1 |
More Information
Set Membership: |
|

Genomic Map