Assay Details
Target Gene Details
Entrez Gene ID: | 3679 |
Gene Name: | integrin subunit alpha 7 |
Gene Aliases: |
- |
Location: |
Chr.12:55684568-55716037 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA7 | NM_001144996.1 | NP_001138468.1 | ||
| NM_001144997.1 | NP_001138469.1 | |||
| NM_002206.2 | NP_002197.2 | |||
| XM_005268839.1 | XP_005268896.1 | |||
| XM_005268840.1 | XP_005268897.1 | |||
| XM_005268841.1 | XP_005268898.1 | |||
| XM_005268842.1 | XP_005268899.1 | |||
| XM_005268844.1 | XP_005268901.1 | |||
| XM_005268846.1 | XP_005268903.1 | |||
| XM_005268848.1 | XP_005268905.1 | |||
| XM_005268849.1 | XP_005268906.1 | |||
| XM_005268850.1 | XP_005268907.1 | |||
| XM_017019265.1 | XP_016874754.1 | |||
| AB210036.1 | ||||
| AF032108.1 | AAC39708.1 | |||
| AK022548.1 | ||||
| AK057949.1 | ||||
| AK304864.1 | ||||
| AY358882.1 | AAQ89241.1 | |||
| BC050280.1 | AAH50280.1 | |||
| X74295.1 | CAA52348.1 |
Target Gene Details
Entrez Gene ID: | 196410 |
Gene Name: | methyltransferase like 7B |
Gene Aliases: |
ALDI |
Location: |
Chr.12:55681546-55684611 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| METTL7B | NM_152637.2 | NP_689850.2 | ||
| AY358508.1 | AAQ88872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525231 | Chr.12:55683135 - 55730548 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map