Assay Details
Target Gene Details
Entrez Gene ID: | 27063 |
Gene Name: | ankyrin repeat domain 1 |
Gene Aliases: |
ALRP, C-193, CARP, CVARP, MCARP, bA320F15.2 |
Location: |
Chr.10:90912100-90921275 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD1 | NM_014391.2 | 9 | 1507 | NP_055206.2 |
| AK290960.1 | ||||
| AY903446.1 | 9 | 1276 | AAX23581.1 | |
| BC018667.1 | 9 | 1323 | AAH18667.1 | |
| X83703.1 | 9 | 1508 | CAA58676.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831945 | Chr.10:90830819 - 91033825 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map