Assay Details
Target Gene Details
Entrez Gene ID: | 284677 |
Gene Name: | chromosome 1 open reading frame 204 |
Gene Aliases: |
- |
Location: |
Chr.1:159834474-159855347 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf204 | NM_001134233.1 | NP_001127705.1 | ||
| AK096506.1 |
Target Gene Details
Entrez Gene ID: | 56833 |
Gene Name: | SLAM family member 8 |
Gene Aliases: |
BLAME, CD353, SBBI42 |
Location: |
Chr.1:159826650-159837497 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLAMF8 | NM_020125.2 | 5 | 2310 | NP_064510.1 |
| XM_005245345.2 | XP_005245402.1 | |||
| XM_011509775.1 | XP_011508077.1 | |||
| XM_011509776.2 | XP_011508078.1 | |||
| XM_011509777.2 | 5 | 3423 | XP_011508079.1 | |
| XM_011509778.2 | XP_011508080.1 | |||
| XM_011509779.2 | XP_011508081.1 | |||
| XM_011509780.2 | 4 | 1917 | XP_011508082.2 | |
| XM_017001844.1 | 6 | 3339 | XP_016857333.1 | |
| XM_017001846.1 | 5 | 2197 | XP_016857335.1 | |
| AF144235.1 | 2 | 1391 | AAD33923.1 | |
| AF146761.1 | 5 | 2188 | AAF67470.1 | |
| AK074669.1 | 5 | 2249 | BAC11123.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005195 | Chr.1:159482969 - 159940547 on Build GRCh38 | Gain |
|
| esv3587703 | Chr.1:159834085 - 159837884 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map