Assay Details
Target Gene Details
Entrez Gene ID: | 10056 |
Gene Name: | phenylalanyl-tRNA synthetase beta subunit |
Gene Aliases: |
FARSLB, FRSB, HSPC173, PheHB, PheRS |
Location: |
Chr.2:222571443-222656355 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FARSB | NM_005687.4 | 17 | 2330 | NP_005678.3 |
| NR_130154.1 | 18 | 2526 | ||
| D84430.1 | 17 | 2061 | BAA95608.1 |
More Information
Set Membership: |
|

Genomic Map