Assay Details
Target Gene Details
Entrez Gene ID: | 256691 |
Gene Name: | MAM domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.9:70043519-70226972 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAMDC2 | NM_153267.4 | 1 | 279 | NP_694999.3 |
| NR_125850.1 | 1 | 279 | ||
| XM_011518471.1 | 1 | 341 | XP_011516773.1 | |
| XM_011518475.2 | 1 | 341 | XP_011516777.1 | |
| XM_017014587.1 | 1 | 341 | XP_016870076.1 | |
| AK304040.1 | ||||
| BC016383.1 | AAH16383.1 | |||
| BC063634.1 | 1 | 256 | AAH63634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831613 | Chr.9:69971142 - 70125145 on Build GRCh38 | Gain |
|

Genomic Map