Assay Details
Target Gene Details
Entrez Gene ID: | 6891 |
Gene Name: | transporter 2, ATP binding cassette subfamily B member |
Gene Aliases: |
ABC18, ABCB3, APT2, D6S217E, PSF-2, PSF2, RING11 |
Location: |
Chr.6:32821833-32838823 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAP2 | NM_018833.2 | 12 | 2282 | NP_061313.2 |
| AF105151.1 | 12 | 2239 | AAD12059.1 | |
| AF152583.1 | 1 | 180 | AAD32715.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1073969 | Chr.6:31456422 - 33480523 on Build GRCh38 | Deletion |
|
| nsv1112900 | Chr.6:31455323 - 33480423 on Build GRCh38 | Deletion |
|
| nsv1119076 | Chr.6:32595823 - 33480623 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map