Assay Details
Target Gene Details
Entrez Gene ID: | 157313 |
Gene Name: | cell division cycle associated 2 |
Gene Aliases: |
PPP1R81, Repo-Man |
Location: |
Chr.8:25458569-25511308 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDCA2 | NM_001317906.1 | 2 | 708 | NP_001304835.1 |
| NM_001317907.1 | NP_001304836.1 | |||
| NM_152562.3 | NP_689775.2 | |||
| XM_006716294.2 | XP_006716357.1 | |||
| XM_011544415.1 | XP_011542717.1 | |||
| XM_011544416.2 | XP_011542718.1 | |||
| XM_011544417.2 | XP_011542719.1 | |||
| XM_011544418.1 | XP_011542720.1 | |||
| AK292505.1 | 2 | 440 | ||
| AK299278.1 | ||||
| AL833627.1 | 2 | 402 | CAH10577.1 | |
| BC104450.1 | AAI04451.1 | |||
| BC104451.1 | AAI04452.1 | |||
| BG716934.1 | ||||
| BM802460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759606 | Chr.8:25411509 - 25564618 on Build GRCh38 | Loss |
|
| dgv7148n100 | Chr.8:25410448 - 25473361 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map