Assay Details
Target Gene Details
Entrez Gene ID: | 80020 |
Gene Name: | FAD dependent oxidoreductase domain containing 2 |
Gene Aliases: |
ERFAD |
Location: |
Chr.22:36487186-36507101 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXRED2 | NM_001102371.1 | 9 | 2832 | NP_001095841.1 |
| NM_024955.5 | 9 | 2871 | NP_079231.4 | |
| XM_017028941.1 | 8 | 2504 | XP_016884430.1 | |
| XM_017028942.1 | 8 | 2601 | XP_016884431.1 | |
| AK025714.1 | 9 | 2771 | BAB15227.1 | |
| AK026975.1 | 6 | 1952 | BAB15610.1 | |
| AK091053.1 | 4 | 1763 | ||
| BC015726.1 | 9 | 2755 | AAH15726.1 | |
| BC027716.2 | 9 | 2850 | AAH27716.1 | |
| BC040351.1 | 9 | 2794 | AAH40351.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760722 | Chr.22:36390921 - 36617980 on Build GRCh38 | Gain |
|
| nsv520497 | Chr.22:36299896 - 36570195 on Build GRCh38 | Gain |
|
| nsv1060004 | Chr.22:36448479 - 36615303 on Build GRCh38 | Gain |
|
| nsv834186 | Chr.22:36335878 - 36510765 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map