Assay Details
Target Gene Details
Entrez Gene ID: | 5871 |
Gene Name: | mitogen-activated protein kinase kinase kinase kinase 2 |
Gene Aliases: |
BL44, GCK, RAB8IP |
Location: |
Chr.11:64786914-64803254 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP4K2 | NM_001307990.1 | 32 | 2535 | NP_001294919.1 |
| NM_004579.4 | 32 | 2559 | NP_004570.2 | |
| XM_011545203.2 | 31 | 2484 | XP_011543505.1 | |
| XM_011545204.2 | 31 | 2421 | XP_011543506.1 | |
| XM_017018093.1 | 31 | 2397 | XP_016873582.1 | |
| XM_017018094.1 | 31 | 2453 | XP_016873583.1 | |
| XM_017018095.1 | 30 | 2384 | XP_016873584.1 | |
| AW574847.1 | 1 | 426 | ||
| BC047865.1 | 32 | 2522 | AAH47865.1 | |
| U07349.1 | 32 | 2490 | AAA20968.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528506 | Chr.11:64577976 - 64933558 on Build GRCh38 | Loss |
|
| dgv72n21 | Chr.11:64778919 - 64817515 on Build GRCh38 | Loss |
|
| nsv469962 | Chr.11:64759608 - 64854101 on Build GRCh38 | Loss |
|
| dgv1965n54 | Chr.11:64566642 - 64806117 on Build GRCh38 | Loss |
|
| nsv951013 | Chr.11:64777329 - 64847128 on Build GRCh38 | Deletion |
|
| dgv153n27 | Chr.11:64709578 - 64817487 on Build GRCh38 | Loss |
|
| nsv832189 | Chr.11:64719711 - 64852727 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map