Assay Details
Target Gene Details
Entrez Gene ID: | 57007 |
Gene Name: | atypical chemokine receptor 3 |
Gene Aliases: |
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1 |
Location: |
Chr.2:236537131-236582358 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACKR3 | NM_020311.2 | 2 | 258 | NP_064707.1 |
| XM_005246097.2 | 2 | 154 | XP_005246154.1 | |
| XM_005246098.3 | 3 | 396 | XP_005246155.1 | |
| XM_017004516.1 | 2 | 149 | XP_016860005.1 | |
| AF030297.1 | 2 | 106 | AAB94130.1 | |
| AK291659.1 | 2 | 170 | ||
| BC036661.2 | 4 | 455 | AAH36661.1 | |
| BM925428.1 | 2 | 202 | ||
| BP426139.1 | 2 | 258 | ||
| DB455412.1 | ||||
| DQ822477.1 | 1 | 44 | ||
| M64749.1 | 1 | 44 | AAA62370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528791 | Chr.2:236576213 - 236581261 on Build GRCh38 | Loss |
|
| nsv1005677 | Chr.2:236369512 - 236811991 on Build GRCh38 | Gain |
|
| nsv524571 | Chr.2:236578945 - 236581261 on Build GRCh38 | Loss |
|
| nsv997438 | Chr.2:236210757 - 237034862 on Build GRCh38 | Gain |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
| nsv834576 | Chr.2:236466567 - 236654210 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map