Assay Details
Target Gene Details
Entrez Gene ID: | 375298 |
Gene Name: | ceramide kinase like |
Gene Aliases: |
RP26 |
Location: |
Chr.2:181536672-181657107 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CERKL | NM_001030311.2 | 14 | 2690 | NP_001025482.1 |
| NM_001030312.2 | 10 | 2273 | NP_001025483.1 | |
| NM_001030313.2 | 11 | 2405 | NP_001025484.1 | |
| NM_001160277.1 | 13 | 2558 | NP_001153749.1 | |
| NM_201548.4 | 13 | 2612 | NP_963842.1 | |
| NR_027689.1 | 11 | 2416 | ||
| NR_027690.1 | 12 | 2548 | ||
| AJ640141.1 | 13 | 2529 | CAG26695.1 | |
| AK129976.1 | 10 | 1855 | BAC85266.1 | |
| BC137498.1 | 13 | 2538 | ||
| BC137499.1 | 13 | 2538 | ||
| BM969366.1 |
Target Gene Details
Entrez Gene ID: | 3676 |
Gene Name: | integrin subunit alpha 4 |
Gene Aliases: |
CD49D, IA4 |
Location: |
Chr.2:181456892-181538928 on Build GRCh38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGA4 | NM_000885.5 | 28 | 5607 | NP_000876.3 |
| AB208885.1 | 25 | 4385 | BAD92122.1 | |
| BC080190.1 | 11 | 2952 | AAH80190.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs116322351] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map