Assay Details
Target Gene Details
Entrez Gene ID: | 4689 |
Gene Name: | neutrophil cytosolic factor 4 |
Gene Aliases: |
CGD3, NCF, P40PHOX, SH3PXD4 |
Location: |
Chr.22:36860988-36878017 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCF4 | NM_000631.4 | NP_000622.2 | ||
| NM_013416.3 | NP_038202.2 | |||
| XM_017028808.1 | XP_016884297.1 | |||
| AB025219.1 | BAA89791.1 | |||
| AB025220.1 | BAA89792.1 | |||
| AK223324.1 | BAD97044.1 | |||
| AK290924.1 | ||||
| BC002798.1 | AAH02798.1 | |||
| BT007346.1 | AAP36010.1 | |||
| BU076892.1 | ||||
| BX355636.2 | ||||
| CR456528.1 | CAG30414.1 | |||
| CR542078.1 | CAG46875.1 | |||
| X77094.1 | CAA54372.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv588983 | Chr.22:36862461 - 36875760 on Build GRCh38 | Loss |
|
| esv2724203 | Chr.22:36872289 - 36872936 on Build GRCh38 | Deletion |
|
| nsv3619 | Chr.22:36856525 - 36877665 on Build GRCh38 | Insertion |
|
| esv2758837 | Chr.22:36779110 - 36964864 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map