Assay Details
Target Gene Details
Entrez Gene ID: | 25998 |
Gene Name: | inhibitor of Bruton tyrosine kinase |
Gene Aliases: |
BTBD26, BTKI |
Location: |
Chr.6:82169987-82248294 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IBTK | NM_001300906.1 | 29 | 5116 | NP_001287835.1 |
| NM_015525.3 | 29 | 5161 | NP_056340.2 | |
| XM_006715453.2 | 30 | 5824 | XP_006715516.1 | |
| XM_006715454.2 | 30 | 5779 | XP_006715517.1 | |
| AB037838.2 | 28 | 4885 | ||
| AF235049.1 | 6 | 1580 | AAG27170.1 | |
| AJ420588.1 | 4 | 1045 | ||
| AJ420593.1 | 5 | 1065 | ||
| AK123460.1 | 2 | 1881 | ||
| AK126777.1 | 13 | 3026 | ||
| AL050018.1 | 9 | 1701 | CAB43239.1 | |
| BC027490.1 | 7 | 1345 | AAH27490.1 | |
| BC038244.2 | 12 | 1999 | AAH38244.2 | |
| BC042171.1 | 8 | 1544 | AAH42171.2 | |
| DQ005633.1 | 29 | 5161 | AAY55906.1 | |
| DQ005635.1 | 5 | 1691 | AAY55908.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830713 | Chr.6:82051484 - 82243853 on Build GRCh38 | Gain |
|
| nsv830715 | Chr.6:82096940 - 82246610 on Build GRCh38 | Loss |
|
| nsv7937 | Chr.6:82130518 - 82535071 on Build GRCh38 | Loss |
|
| nsv470837 | Chr.6:82147657 - 82223592 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map